2-(3-Nitrobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086612-1G
086612-2G
086612-5G
102849-586EA
795.38
USD
102849-586
102849-588
102849-590
2-(3-Nitrobenzoyl)pyridine ≥97%
2-(3-Nitrobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₈N₂O₃ |
MDL Number:
MFCD00957342 CAS Number: 27693-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...