2-(4-Cyanobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086610-1G
086610-2G
086610-5G
102849-574EA
795.38
USD
102849-574
102849-576
102849-578
2-(4-Cyanobenzoyl)pyridine ≥97%
2-(4-Cyanobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₈N₂O |
MDL Number:
MFCD07699243 CAS Number: 57954-94-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...