2-(3-Cyanobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086609-1G 086609-2G 086609-5G
102849-568EA 795.38 USD
102849-568 102849-570 102849-572
2-(3-Cyanobenzoyl)pyridine ≥97%
2-(3-Cyanobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₃H₈N₂O
MDL Number: MFCD07699242
CAS Number: 898779-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR