2-(4-Bromobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086607-1G 086607-2G 086607-5G
102849-556EA 766.15 USD
102849-556 102849-558 102849-560
2-(4-Bromobenzoyl)pyridine ≥97%
2-(4-Bromobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈BrNO
Boiling Pt: 369 °C
Melting Pt: 46…48 °C
Density: 1.48
Flash Pt: 177 °C
MDL Number: MFCD07699238
CAS Number: 18453-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR