2-(3-Bromobenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086606-1G
086606-2G
086606-5G
102849-550EA
795.38
USD
102849-550
102849-552
102849-554
2-(3-Bromobenzoyl)pyridine ≥97%
2-(3-Bromobenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₂H₈BrNO MW: 262.11 g/mol |
MDL Number:
MFCD07699237 CAS Number: 206357-82-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...