2-(2-Bromobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086605-1G 086605-2G 086605-5G
102849-544EA 795.38 USD
102849-544 102849-546 102849-548
2-(2-Bromobenzoyl)pyridine ≥97%
2-(2-Bromobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈BrNO
MDL Number: MFCD07699236
CAS Number: 76160-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR