2-(4-Fluorobenzoyl)pyridine ≥97%

Supplier: Matrix Scientific

086603-1G 086603-2G 086603-5G
102849-532EA 1158.46 USD
102849-532 102849-534 102849-536
2-(4-Fluorobenzoyl)pyridine ≥97%
2-(4-Fluorobenzoyl)pyridine

Hazard: Irritant

Formula: C₁₂H₈FNO
MDL Number: MFCD07699233
CAS Number: 169955-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR