2-(4-Methoxybenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086598-1G
086598-2G
086598-5G
102849-502EA
795.38
USD
102849-502
102849-504
102849-506
2-(4-Methoxybenzoyl)pyridine ≥97%
2-(4-Methoxybenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ |
MDL Number:
MFCD01716432 CAS Number: 6305-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...