2-(3-Methoxybenzoyl)pyridine ≥97%
Supplier: Matrix Scientific
086597-1G
086597-2G
086597-5G
102849-496EA
795.38
USD
102849-496
102849-498
102849-500
2-(3-Methoxybenzoyl)pyridine ≥97%
2-(3-Methoxybenzoyl)pyridine
Hazard: Irritant
Formula:
C₁₃H₁₁NO₂ |
MDL Number:
MFCD07699224 CAS Number: 55030-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...