t-Butyl 2-pyridyl ketone ≥97%

Supplier: Matrix Scientific

086594-1G 086594-2G 086594-5G
102849-478EA 795.38 USD
102849-478 102849-480 102849-482
t-Butyl 2-pyridyl ketone ≥97%
t-Butyl 2-pyridyl ketone

Hazard: Irritant

Formula: C₁₀H₁₃NO
MDL Number: MFCD06738273
CAS Number: 31595-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR