Cyclohexyl-2-pyridyl ketone ≥97%
Supplier: Matrix Scientific
086592-1G
086592-2G
086592-5G
102849-466EA
795.38
USD
102849-466
102849-468
102849-470
Cyclohexyl-2-pyridyl ketone ≥97%
Cyclohexyl-2-pyridyl ketone
Hazard: Irritant
Formula:
C₁₂H₁₅NO |
MDL Number:
MFCD00778683 CAS Number: 6602-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...