Cyclopentyl-2-pyridyl ketone ≥97%
Supplier: Matrix Scientific
086591-1G
086591-2G
086591-5G
102849-460EA
795.38
USD
102849-460
102849-462
102849-464
Cyclopentyl-2-pyridyl ketone ≥97%
Cyclopentyl-2-pyridyl ketone
Hazard: Irritant
Formula:
C₁₁H₁₃NO |
MDL Number:
MFCD07699202 CAS Number: 157592-43-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...