Cyclobutyl-2-pyridyl ketone ≥97%

Supplier: Matrix Scientific

086590-1G 086590-2G 086590-5G
102849-454EA 795.38 USD
102849-454 102849-456 102849-458
Cyclobutyl-2-pyridyl ketone ≥97%
Cyclobutyl-2-pyridyl ketone

Hazard: Irritant

Formula: C₁₀H₁₁NO
MDL Number: MFCD07699201
CAS Number: 515154-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR