Cyclopentyl-3-thienyl ketone ≥97%
Supplier: Matrix Scientific
086585-1G
086585-2G
086585-5G
102849-424EA
795.38
USD
102849-424
102849-426
102849-428
Cyclopentyl-3-thienyl ketone ≥97%
Cyclopentyl-3-thienyl ketone
Hazard: Irritant
Formula:
C₁₀H₁₂OS |
MDL Number:
MFCD07699056 CAS Number: 898771-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...