Cyclobutyl-3-thienyl ketone ≥97%
Supplier: Matrix Scientific
086584-1G
086584-2G
086584-5G
102849-418EA
795.38
USD
102849-418
102849-420
102849-422
Cyclobutyl-3-thienyl ketone ≥97%
Cyclobutyl-3-thienyl ketone
Hazard: Irritant
Formula:
C₉H₁₀OS |
MDL Number:
MFCD07699055 CAS Number: 898771-70-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...