Cyclopropyl-3-thienyl ketone ≥97%
Supplier: Matrix Scientific
086583-1G
086583-2G
086583-5G
102849-412EA
795.38
USD
102849-412
102849-414
102849-416
Cyclopropyl-3-thienyl ketone ≥97%
Cyclopropyl-3-thienyl ketone
Hazard: Irritant
Formula:
C₈H₈OS |
MDL Number:
MFCD07699054 CAS Number: 113348-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...