3-(4-Bromo-3-fluorobenzoyl)thiophene ≥97%

Supplier: Matrix Scientific

086580-1G 086580-2G 086580-5G
102849-394EA 795.38 USD
102849-394 102849-396 102849-398
3-(4-Bromo-3-fluorobenzoyl)thiophene ≥97%
3-(4-Bromo-3-fluorobenzoyl)thiophene

Hazard: Irritant

Formula: C₁₁H₆BrFOS
MDL Number: MFCD07699032
CAS Number: 898771-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR