Ethyl-5-(3-oxocyclohexyl)-2-furoate ≥97%

Supplier: Matrix Scientific

086577-1G 086577-2G 086577-5G
102849-376EA 795.38 USD
102849-376 102849-378 102849-380
Ethyl-5-(3-oxocyclohexyl)-2-furoate ≥97%
Ethyl-5-(3-oxocyclohexyl)-2-furoate

Hazard: Irritant

Formula: C₁₃H₁₆O₄
MDL Number: MFCD03841030
CAS Number: 883898-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR