Ethyl-5-(3-oxocyclohexyl)-2-thiophenecarboxylate ≥97%

Supplier: Matrix Scientific

086575-1G 086575-2G 086575-5G
102849-364EA 795.38 USD
102849-364 102849-366 102849-368
Ethyl-5-(3-oxocyclohexyl)-2-thiophenecarboxylate ≥97%
Ethyl-5-(3-oxocyclohexyl)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₃H₁₆O₃S
MDL Number: MFCD03841026
CAS Number: 909421-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR