(2-Chloropyridin-5-yl)thiazolyl ketone ≥97%

Supplier: Matrix Scientific

086568-1G 086568-2G 086568-5G
102849-322EA 795.38 USD
102849-322 102849-324 102849-326
(2-Chloropyridin-5-yl)thiazolyl ketone ≥97%
(2-Chloropyridin-5-yl)thiazolyl ketone

Hazard: Irritant

Formula: C₉H₅ClN₂OS
MDL Number: MFCD07699005
CAS Number: 890100-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR