2-Chloro-3',4'-(ethylenedioxy)benzophenone ≥97%

Supplier: Matrix Scientific

086563-1G 086563-2G 086563-5G
102849-292EA 490.77 USD
102849-292 102849-294 102849-296
2-Chloro-3',4'-(ethylenedioxy)benzophenone ≥97%
2-Chloro-3',4'-(ethylenedioxy)benzophenone

Hazard: Irritant

Formula: C₁₅H₁₁ClO₃
MDL Number: MFCD00604246
CAS Number: 164526-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR