4-Chloro-3',4'-dimethoxybenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Chlorophenyl)(3,4-dimethoxyphenyl)methanone

086562-1G 086562-2G 086562-5G
102849-286EA 490.77 USD
102849-286 102849-288 102849-290
4-Chloro-3',4'-dimethoxybenzophenone ≥97%
4-Chloro-3',4'-dimethoxybenzophenone

Hazard: Irritant

Formula: C₁₅H₁₃ClO₃
MW: 276.7149 g/mol
MDL Number: MFCD02127307
CAS Number: 116412-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR