3,3',5,5'-Tetrafluorobenzophenone ≥97%
Supplier: Matrix Scientific
086561-1G
086561-2G
086561-5G
102849-280EA
489.23
USD
102849-280
102849-282
102849-284
3,3',5,5'-Tetrafluorobenzophenone ≥97%
3,3',5,5'-Tetrafluorobenzophenone
Hazard: Irritant
Formula:
C₁₃H₆F₄O Melting Pt: 85-87°C |
MDL Number:
MFCD06201523 CAS Number: 139911-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...