3,3',5,5'-Tetrachlorobenzophenone ≥97%
Supplier: Matrix Scientific
086560-1G
086560-2G
086560-5G
102849-274EA
489.23
USD
102849-274
102849-276
102849-278
3,3',5,5'-Tetrachlorobenzophenone ≥97%
3,3',5,5'-Tetrachlorobenzophenone
Hazard: Irritant
Formula:
C₁₃H₆Cl₄O Melting Pt: 161…164 °C |
MDL Number:
MFCD01763607 CAS Number: 92758-86-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...