2-Bromo-4'-chlorobenzophenone ≥97%
Supplier: Matrix Scientific
086559-1G
086559-2G
086559-5G
102849-268EA
673.85
USD
102849-268
102849-270
102849-272
2-Bromo-4'-chlorobenzophenone ≥97%
2-Bromo-4'-chlorobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈BrClO |
MDL Number:
MFCD08063806 CAS Number: 99585-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...