4-Acetoxy-3',4'-dichlorobenzophenone ≥90%
Supplier: Matrix Scientific
086558-1G
086558-2G
086558-5G
102849-262EA
489.23
USD
102849-262
102849-264
102849-266
4-Acetoxy-3',4'-dichlorobenzophenone ≥90%
4-Acetoxy-3',4'-dichlorobenzophenone
Hazard: Irritant
Formula:
C₁₅H₁₀Cl₂O₃ Melting Pt: 105…107 °C |
MDL Number:
MFCD07698987 CAS Number: 890100-13-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...