4-Acetoxy-4'-nitrobenzophenone ≥90%

Supplier: Matrix Scientific

086557-1G 086557-2G 086557-5G
102849-256EA 490.77 USD
102849-256 102849-258 102849-260
4-Acetoxy-4'-nitrobenzophenone ≥90%
4-Acetoxy-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₅H₁₁NO₅
Melting Pt: 130…131 °C
MDL Number: MFCD07698961
CAS Number: 890099-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR