4-Acetoxy-4'-iodobenzophenone ≥90%

Supplier: Matrix Scientific

086556-1G 086556-2G 086556-5G
102849-250EA 490.77 USD
102849-250 102849-252 102849-254
4-Acetoxy-4'-iodobenzophenone ≥90%
4-Acetoxy-4'-iodobenzophenone

Hazard: Irritant

Formula: C₁₅H₁₁IO₃
MDL Number: MFCD07698955
CAS Number: 890099-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR