4-Acetoxy-4'-chlorobenzophenone ≥90%
Supplier: Matrix Scientific
086555-1G
086555-2G
086555-5G
102849-244EA
490.77
USD
102849-244
102849-246
102849-248
4-Acetoxy-4'-chlorobenzophenone ≥90%
4-Acetoxy-4'-chlorobenzophenone
Hazard: Irritant
Formula:
C₁₅H₁₁ClO₃ |
MDL Number:
MFCD07698949 CAS Number: 103060-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...