4-Acetoxy-4'-fluorobenzophenone ≥90%
Supplier: Matrix Scientific
086554-1G
086554-2G
086554-5G
102849-238EA
490.77
USD
102849-238
102849-240
102849-242
4-Acetoxy-4'-fluorobenzophenone ≥90%
4-Acetoxy-4'-fluorobenzophenone
Hazard: Irritant
Formula:
C₁₅H₁₁FO₃ |
MDL Number:
MFCD07698946 CAS Number: 133830-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...