4-Cyano-4'-nitrobenzophenone ≥97%

Supplier: Matrix Scientific

086551-1G 086551-2G 086551-5G
102849-220EA 710.77 USD
102849-220 102849-222 102849-224
4-Cyano-4'-nitrobenzophenone ≥97%
4-Cyano-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₄H₈N₂O₃
Melting Pt: 151…153 °C
MDL Number: MFCD03841021
CAS Number: 68271-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR