4-Fluoro-3-methyl-4'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
086550-1G
086550-2G
086550-5G
102849-214EA
490.77
USD
102849-214
102849-216
102849-218
4-Fluoro-3-methyl-4'-nitrobenzophenone ≥97%
4-Fluoro-3-methyl-4'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₀FNO₃ |
MDL Number:
MFCD03841022 CAS Number: 760192-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...