4-Nitro-4'-thiomethylbenzophenone ≥97%
Supplier: Matrix Scientific
086549-1G
086549-2G
086549-5G
102849-208EA
490.77
USD
102849-208
102849-210
102849-212
4-Nitro-4'-thiomethylbenzophenone ≥97%
4-Nitro-4'-thiomethylbenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁NO₃S |
MDL Number:
MFCD07698795 CAS Number: 127615-48-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...