1-Naphthyl-4-nitrophenyl ketone ≥97%
Supplier: Matrix Scientific
086548-1G
086548-2G
086548-5G
102849-202EA
383.08
USD
102849-202
102849-204
102849-206
1-Naphthyl-4-nitrophenyl ketone ≥97%
1-Naphthyl-4-nitrophenyl ketone
Hazard: Irritant
Formula:
C₁₇H₁₁NO₃ |
MDL Number:
MFCD03841018 CAS Number: 42495-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...