4-Cyano-4'-iodobenzophenone ≥97%

Supplier: Matrix Scientific

086547-1G 086547-2G 086547-5G
102849-196EA 490.77 USD
102849-196 102849-198 102849-200
4-Cyano-4'-iodobenzophenone ≥97%
4-Cyano-4'-iodobenzophenone

Hazard: Irritant

Formula: C₁₄H₈INO
MDL Number: MFCD02260430
CAS Number: 890098-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR