4-Ethoxycarbonyl-4'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
086545-1G
086545-2G
086545-5G
102849-184EA
710.77
USD
102849-184
102849-186
102849-188
4-Ethoxycarbonyl-4'-nitrobenzophenone ≥97%
4-Ethoxycarbonyl-4'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₆H₁₃NO₅ Melting Pt: 106…108 °C |
MDL Number:
MFCD02260409 CAS Number: 760192-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...