2-Ethoxycarbonyl-4'-nitrobenzophenone ≥97%

Supplier: Matrix Scientific

086544-1G 086544-2G 086544-5G
102849-178EA 489.23 USD
102849-178 102849-180 102849-182
2-Ethoxycarbonyl-4'-nitrobenzophenone ≥97%
2-Ethoxycarbonyl-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₆H₁₃NO₅
Melting Pt: 106…108 °C
MDL Number: MFCD02260407
CAS Number: 760192-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR