4-Fluoro-3'-iodobenzophenone ≥97%

Supplier: Matrix Scientific

086543-1G 086543-2G 086543-5G
102849-172EA 387.69 USD
102849-172 102849-174 102849-176
4-Fluoro-3'-iodobenzophenone ≥97%
4-Fluoro-3'-iodobenzophenone

Hazard: Irritant

Formula: C₁₃H₈FIO
MDL Number: MFCD02260397
CAS Number: 395-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR