4-Bromo-4'-nitrobenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Bromophenyl)(4-nitrophenyl)methanone

086542-1G 086542-2G 086542-5G
102849-166EA 300 USD
102849-166 102849-168 102849-170
4-Bromo-4'-nitrobenzophenone ≥97%
4-Bromo-4'-nitrobenzophenone

Hazard: Irritant

Formula: C₁₃H₈BrNO₃
MW: 306.12 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD00179024
CAS Number: 40292-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR