3-Bromo-4'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
086541-1G
086541-2G
086541-5G
102849-160EA
387.69
USD
102849-160
102849-162
102849-164
3-Bromo-4'-nitrobenzophenone ≥97%
3-Bromo-4'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈BrNO₃ Melting Pt: 114…118 °C |
MDL Number:
MFCD02260388 CAS Number: 760192-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...