4-Chloro-4'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
(4-Chlorophenyl)(4-nitrophenyl)methanone
086539-1G
086539-2G
086539-5G
102849-148EA
387.69
USD
102849-148
102849-150
102849-152
4-Chloro-4'-nitrobenzophenone ≥97%
4-Chloro-4'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈ClNO₃ MW: 261.6605 g/mol |
MDL Number:
MFCD00047743 CAS Number: 7497-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...