3-Chloro-3'-nitrobenzophenone ≥97%
Supplier: Matrix Scientific
086537-1G
086537-2G
086537-5G
102849-136EA
490.77
USD
102849-136
102849-138
102849-140
3-Chloro-3'-nitrobenzophenone ≥97%
3-Chloro-3'-nitrobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈ClNO₃ |
MDL Number:
MFCD00228117 CAS Number: 494203-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...