4-Chloro-4'-iodobenzophenone ≥97%

Supplier: Matrix Scientific

086536-1G 086536-2G 086536-5G
102849-130EA 292.31 USD
102849-130 102849-132 102849-134
4-Chloro-4'-iodobenzophenone ≥97%
4-Chloro-4'-iodobenzophenone

Hazard: Irritant

Formula: C₁₃H₈ClIO
Melting Pt: 167…169 °C
MDL Number: MFCD02260373
CAS Number: 99847-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR