4'-Chloro-2-iodobenzophenone ≥97%
Supplier: Matrix Scientific
086534-1G
086534-2G
086534-5G
102849-118EA
490.77
USD
102849-118
102849-120
102849-122
4'-Chloro-2-iodobenzophenone ≥97%
4'-Chloro-2-iodobenzophenone
Hazard: Irritant
Formula:
C₁₃H₈ClIO |
MDL Number:
MFCD02260367 CAS Number: 99847-66-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...