3-Bromo-4'-methoxybenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (3-Bromophenyl)(4-methoxyphenyl)methanone

086527-1G 086527-2G 086527-5G
102849-076EA 113.85 USD
102849-076 102849-078 102849-080
3-Bromo-4'-methoxybenzophenone ≥97%
3-Bromo-4'-methoxybenzophenone

Hazard: Irritant/Refrigerate

Formula: C₁₄H₁₁BrO₂
MW: 291.139 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD01311588
CAS Number: 54118-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR