2-Carboethoxy-4'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086525-1G
086525-2G
086525-5G
102849-064EA
490.77
USD
102849-064
102849-066
102849-068
2-Carboethoxy-4'-methoxybenzophenone ≥97%
2-Carboethoxy-4'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₇H₁₆O₄ |
MDL Number:
MFCD01311576 CAS Number: 51432-00-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...