3-Cyano-4'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086523-1G
086523-2G
086523-5G
102849-052EA
489.23
USD
102849-052
102849-054
102849-056
3-Cyano-4'-methoxybenzophenone ≥97%
3-Cyano-4'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₅H₁₁NO₂ Melting Pt: 94…95 °C |
MDL Number:
MFCD01311593 CAS Number: 60694-67-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...