4-Chloro-3'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086522-1G
086522-2G
086522-5G
102849-046EA
490.77
USD
102849-046
102849-048
102849-050
4-Chloro-3'-methoxybenzophenone ≥97%
4-Chloro-3'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁ClO₂ |
MDL Number:
MFCD01311567 CAS Number: 32363-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...