2-Carboethoxy-3'-methoxybenzophenone ≥97%

Supplier: Matrix Scientific

086521-1G 086521-2G 086521-5G
102849-040EA 489.23 USD
102849-040 102849-042 102849-044
2-Carboethoxy-3'-methoxybenzophenone ≥97%
2-Carboethoxy-3'-methoxybenzophenone

Hazard: Irritant

Formula: C₁₇H₁₆O₄
Melting Pt: 59…70 °C
MDL Number: MFCD01311547
CAS Number: 750633-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR