3-Methoxybenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (3-Methoxyphenyl)(phenyl)methanone

086519-1G 086519-2G 086519-5G
102849-028EA 490.77 USD
102849-028 102849-030 102849-032
3-Methoxybenzophenone ≥97%
3-Methoxybenzophenone

Hazard: Irritant

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Melting Pt: 36-38°C
MDL Number: MFCD01319936
CAS Number: 6136-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR